tert-Butoxydiphenylchlorosilane ≥95.0% stabilized

Supplier: TCI America

Synonyms: tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride

B1436-5ML B1436-25ML
TCB1436-005MLEA 209 USD
TCB1436-005ML TCB1436-25ML
tert-Butoxydiphenylchlorosilane ≥95.0% stabilized
tert-Butoxydiphenylchlorosilane

(stabilized with CaCO3)
CAS Number: 17922-24-6
MDL Number: MFCD00077672
Molecular Formula: C16H19ClOSi
Molecular Weight: 290.86
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 56
Specific Gravity (20/20): 1.08
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride

Formula: C₁₆H₁₉ClOSi
MW: 290.86 g/mol
Boiling Pt: 112 °C
Density: 1.08 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00077672
CAS Number: 17922-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fb051e7eb88d4b2cfd2c6267735d6ceec3bfc62&vid=7fb051e7eb88d4b2cfd2c6267735d6ceec3bfc62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR