N-(tert-Butoxycarbonyl)-L-phenylalanine ≥99.0%
Supplier: TCI America
Synonyms:
N-Boc-L-phenylalanine, BOC-L-Phenylalanine, N-α-t-Boc-L-phenylalanine, Boc-Phe-OH
B1332-5G
B1332-25G
TCB1332-005GEA
16.62
USD
TCB1332-005G
TCB1332-025G
N-(tert-Butoxycarbonyl)-L-phenylalanine ≥99.0%
N-(tert-Butoxycarbonyl)-L-phenylalanine
CAS Number: 13734-34-4
MDL Number: MFCD00002663
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: 24.5 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-phenylalanine, Boc-Phe-OH
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD00002663 CAS Number: 13734-34-4 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...