Bis(2-dimethylaminoethyl)ether ≥98.0%

Supplier: TCI America

Synonyms: 2-Dimethylaminoethyl Ether, 2,2'-Oxybis(N,N-dimethylethanamine)

B1291-500ML B1291-25ML
TCB1291-500MLEA 174.7 USD
TCB1291-500ML TCB1291-25ML
Bis(2-dimethylaminoethyl)ether ≥98.0%
Bis(2-dimethylaminoethyl)ether

CAS Number: 3033-62-3
MDL Number: MFCD00059199
Molecular Formula: C8H20N2O
Molecular Weight: 160.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 56
Specific Gravity (20/20): 0.85


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: 2-Dimethylaminoethyl Ether

Formula: C₈H₂₀N₂O
Boiling Pt: 80 °C
Density: 0.85 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00059199
CAS Number: 3033-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bb8864a9cf5239aacd5a997dec175576342bece&vid=3bb8864a9cf5239aacd5a997dec175576342bece&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR