4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%

Supplier: TCI America

Synonyms: 4-(tert-butyl)cyclohexanecarboxylic acid

B1285-5G B1285-25G
TCB1285-5GEA 112 USD
TCB1285-5G TCB1285-025G
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 5451-55-8
MDL Number: MFCD00042622
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₁₁H₂₀O₂
MDL Number: MFCD00042622
CAS Number: 5451-55-8

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR