2-(2-Bromoethyl)-1,3-dioxane ≥97.0%
Supplier: TCI America
CAS Number: 33884-43-4
MDL Number: MFCD00006567
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 97
Specific Gravity (20/20): 1.44
Other Product Names / Synonyms: 3-Bromopropionaldehyde Trimethylene Acetal
Formula:
C₆H₁₁BrO₂ Boiling Pt: 70 °C Density: 1.44 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00006567 CAS Number: 33884-43-4 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...