Benzyltrimethylammonium hydroxide 40% in water
Supplier: TCI America
Synonyms:
Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
B1071-500ML
B1071-25ML
TCB1071-500MLEA
288
USD
TCB1071-500ML
TCB1071-25ML
Benzyltrimethylammonium hydroxide 40% in water
Benzyltrimethylammonium hydroxide
CAS Number: 100-85-6
MDL Number: MFCD00008281
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.07
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Density: 1.07 (20 °C) |
MDL Number:
MFCD00008281 CAS Number: 100-85-6 UN: 3286 ADR: 3,I |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...