O-Benzyl-DL-serine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-DL-OBzl-Ser-OH, H-DL-Ser(Bzl)-OH
B0860-1G
B0860-10G
TCB0860-001GEA
25
USD
TCB0860-001G
TCB0860-010G
O-Benzyl-DL-serine ≥98.0% (by titrimetric analysis)
O-Benzyl-DL-serine
CAS Number: 5445-44-3
MDL Number: MFCD00021724
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-DL-Ser(Bzl)-OH
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol |
MDL Number:
MFCD00021724 CAS Number: 5445-44-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...