2-Bromoisobutyryl bromide ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-2-methylpropionyl bromide

B0607-500G B0607-25G
TCB0607-500GEA 184.6 USD
TCB0607-500G TCB0607-25G
2-Bromoisobutyryl bromide ≥98.0%
2-Bromoisobutyryl bromide

CAS Number: 20769-85-1
MDL Number: MFCD00000122
Molecular Formula: C4H6Br2O
Molecular Weight: 229.90
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 110
Specific Gravity (20/20): 1.87


Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 2-Bromo-2-methylpropionyl Bromide

Formula: C₄H₆Br₂O
MW: 229.91 g/mol
Boiling Pt: 164 °C
Density: 1.87 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000122
CAS Number: 20769-85-1
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac6ce2af825cfc94b93644db571ddf6f6a366d3&vid=1ac6ce2af825cfc94b93644db571ddf6f6a366d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR