Bis(cyclohexanone)oxalyldihydrazone ≥98.0%

Supplier: TCI America

Synonyms: Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone

B0476-25G B0476-1G
TCB0476-025GEA 193 USD
TCB0476-025G TCB0476-001G
Bis(cyclohexanone)oxalyldihydrazone ≥98.0%
Bis(cyclohexanone)oxalyldihydrazone

CAS Number: 370-81-0
MDL Number: MFCD00001659
Molecular Formula: C14H22N4O2
Molecular Weight: 278.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 208


Other Product Names / Synonyms: Cupferazone, Cuprizone

Formula: C₁₄H₂₂N₄O₂
MW: 278.36 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00001659
CAS Number: 370-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d400551978c2c5bf09f0ca90ddc2e6b9d4a01faf&vid=d400551978c2c5bf09f0ca90ddc2e6b9d4a01faf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR