Phenylphosphinic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

B0025-500G B0025-25G
TCB0025-500GEA 134 USD
TCB0025-500G TCB0025-25G
Phenylphosphinic acid ≥99.0% (by titrimetric analysis)
Phenylphosphinic acid

CAS Number: 1779-48-2
MDL Number: MFCD00002131
Molecular Formula: C6H7O2P
Molecular Weight: 142.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 85


Other Product Names / Synonyms: Benzenephosphinic Acid, Benzenephosphonous Acid, Phenylphosphonous Acid

Formula: C₆H₇O₂P
MW: 142.092 g/mol
Boiling Pt: 180 °C
Melting Pt: 85 °C
MDL Number: MFCD00002131
CAS Number: 1779-48-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b427485760bda6f036f6660844936d75df2a7357&vid=b427485760bda6f036f6660844936d75df2a7357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR