1-Aminopiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2241-25G A2241-5G
TCA2241-25GEA 156 USD
TCA2241-25G TCA2241-5G
1-Aminopiperidine ≥98.0% (by GC, titration analysis)
1-Aminopiperidine

CAS Number: 2213-43-6
MDL Number: MFCD00006489
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 36
Specific Gravity (20/20): 0.93

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 146 °C
Density: 0.93 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00006489
CAS Number: 2213-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27865d3e79444ee12815597215e583073b652d1c&vid=27865d3e79444ee12815597215e583073b652d1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR