7-Aminodesacetoxycephalosporanic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Aminodeacetoxycephalosporanic acid, 7-Amino-3-methyl-3-cephem-4-carboxylic acid, 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA

A2075-25G A2075-5G
TCA2075-25GEA 109 USD
TCA2075-25G TCA2075-5G
7-Aminodesacetoxycephalosporanic acid ≥98.0% (by HPLC)
7-Aminodesacetoxycephalosporanic acid

CAS Number: 22252-43-3
MDL Number: MFCD00151456
Molecular Formula: C8H10N2O3S
Molecular Weight: 214.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 242
Specific rotation [a]20/D: 104 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 7-ADCA

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00151456
CAS Number: 22252-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=807f07819a18bae79605cfe6ac0461e5936e5a9b&vid=807f07819a18bae79605cfe6ac0461e5936e5a9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR