4-Aminoindole ≥98.0%
Supplier: TCI America
Synonyms:
Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole
A2070-1G
A2070-5G
TCA2070-1GEA
70
USD
TCA2070-1G
TCA2070-5G
4-Aminoindole ≥98.0%
4-Aminoindole
CAS Number: 5192-23-4
MDL Number: MFCD01076559
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 109
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 108 |
MDL Number:
MFCD01076559 CAS Number: 5192-23-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...