4-Amino-m-anisic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

A2029-25G A2029-5G
TCA2029-25GEA 399 USD
TCA2029-25G TCA2029-5G
4-Amino-m-anisic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-m-anisic acid

CAS Number: 2486-69-3
MDL Number: MFCD00016539
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 184


Other Product Names / Synonyms: 4-Amino-m-anisic Acid, o-Anisidine-4-carboxylic Acid

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00016539
CAS Number: 2486-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=921d32d167572ccf12f702a075efa0f361ce2cd7&vid=921d32d167572ccf12f702a075efa0f361ce2cd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR