Albendazole ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
[5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester, Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate
A1943-5G
A1943-25G
TCA1943-5GEA
37.65
USD
TCA1943-5G
TCA1943-25G
Albendazole ≥98.0% (by total nitrogen basis)
Albendazole
CAS Number: 54965-21-8
MDL Number: MFCD00083232
Molecular Formula: C12H15N3O2S
Molecular Weight: 265.33
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 210
Product Categories: Bioscience, Anthelmintic Ingredients
Other Product Names / Synonyms: Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate, [5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester
Formula:
C₁₂H₁₅N₃O₂S MW: 265.33 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00083232 CAS Number: 54965-21-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...