2,6-Dichloro-4-(trifluoromethyl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-3,5-dichloro-α,α,α-trifluorotoluene, 4-Amino-3,5-dichlorobenzotrifluoride, 2,6-Dichloro-4-trifluoromethylaniline, 2,6-Dichloro-4-(trifluoromethyl)aniline
CAS Number: 24279-39-8
MDL Number: MFCD00052918
Molecular Formula: C7H4Cl2F3N
Molecular Weight: 230.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 62
Melting point (°C): 37
Flash Point (°C): 88
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Dichloro-4-(trifluoromethyl)aniline
Formula:
C₇H₄Cl₂F₃N MW: 230.02 Boiling Pt: 62 °C Melting Pt: 37 °C Density: 1.532 Flash Pt: 88 °C |
MDL Number:
MFCD00052918 CAS Number: 24279-39-8 UN: 3077 ADR: 9 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...