2-Methoxyethyl acrylate ≥98.0% stabilised

Supplier: TCI America

Synonyms: Ethylene glycol methyl ether acrylate, Ethylene Glycol Monomethyl Ether Acrylate, (2-Methoxyethyl) acrylate, Acrylic Acid 2-Methoxyethyl Ester

A1405-25ML A1405-500ML
TCA1405-025MLEA 17 USD
TCA1405-025ML TCA1405-500ML
2-Methoxyethyl acrylate ≥98.0% stabilised
2-Methoxyethyl acrylate

(stabilized with MEHQ)
CAS Number: 3121-61-7
MDL Number: MFCD00048149
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Flash Point (°C): 60
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2-Methoxyethyl Ester, Ethylene Glycol Monomethyl Ether Acrylate

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 56 °C
Density: 1.01 (20 °C)
Flash Pt: 60°C
MDL Number: MFCD00048149
CAS Number: 3121-61-7
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=109e58052947a9e2af30bb2297cb9a118692052f&vid=109e58052947a9e2af30bb2297cb9a118692052f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR