2-Phenoxyethyl acrylate ≥93.0% stabilized

Supplier: TCI America

Synonyms: Acrylic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acrylate

A1400-25G A1400-500G
TCA1400-025GEA 38.67 USD
TCA1400-025G TCA1400-500G
2-Phenoxyethyl acrylate ≥93.0% stabilized
2-Phenoxyethyl acrylate

(stabilized with MEHQ)
CAS Number: 48145-04-6
MDL Number: MFCD00053696
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Flash Point (°C): 141
Specific Gravity (20/20): 1.11


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Ethylene Glycol Monophenyl Ether Acrylate, Acrylic Acid 2-Phenoxyethyl Ester

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Density: 1.11 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00053696
CAS Number: 48145-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32743ada89d39a5ca6fa9561afe055cd759da09c&vid=32743ada89d39a5ca6fa9561afe055cd759da09c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR