tert-Butyl acrylate ≥98.0% stabilized
Supplier: TCI America
Synonyms:
Acrylic Acid tert-Butyl Ester
A1389-500ML
A1389-25ML
TCA1389-500MLEA
85.45
USD
TCA1389-500ML
TCA1389-25ML
tert-Butyl acrylate ≥98.0% stabilized
tert-Butyl acrylate
(stabilized with MEHQ)
CAS Number: 1663-39-4
MDL Number: MFCD00008809
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 19
Specific Gravity (20/20): 0.88
Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: Acrylic Acid tert-Butyl Ester
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 127 °C Density: 0.88 (20 °C) Flash Pt: 19 °C |
MDL Number:
MFCD00008809 CAS Number: 1663-39-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...