Ethyl-3-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl
CAS Number: 4244-84-2
MDL Number: MFCD00012909
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 69
Other Product Names / Synonyms: 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-Aminopropionate Hydrochloride
Formula:
C₅H₁₁NO₂·HCl MW: 153.6 g/mol Melting Pt: 69 °C |
MDL Number:
MFCD00012909 CAS Number: 4244-84-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...