Hexyl acrylate ≥96.0% stabilized
Supplier: TCI America
(stabilized with HQ)
CAS Number: 2499-95-8
MDL Number: MFCD00048881
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 68
Specific Gravity (20/20): 0.89
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Hexyl Ester
Formula:
C₉H₁₆O₂ Boiling Pt: 89 °C Density: 0.89 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00048881 CAS Number: 2499-95-8 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...