2-(Dimethylamino)ethyl acrylate ≥98.0% stabilized
Supplier: TCI America
Synonyms:
Acrylic Acid 2-(Dimethylamino)ethyl Ester
A1235-500G
A1235-25G
A1235-100G
TCA1235-500GEA
194.59
USD
TCA1235-500G
TCA1235-25G
TCA1235-100G
2-(Dimethylamino)ethyl acrylate ≥98.0% stabilized
2-(Dimethylamino)ethyl acrylate
(stabilized with MEHQ)
CAS Number: 2439-35-2
MDL Number: MFCD00038233
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 59
Specific Gravity (20/20): 0.94
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Acrylic Acid 2-(Dimethylamino)ethyl Ester
Formula:
C₇H₁₃NO₂ MW: 143.19 g/mol Boiling Pt: 161 °C Density: 0.94 (20 °C) Flash Pt: 59 °C |
MDL Number:
MFCD00038233 CAS Number: 2439-35-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...