Cyclohexyl acrylate ≥98.0% stabilized

Supplier: TCI America

A1208-500ML A1208-25ML
TCA1208-500MLEA 170.87 USD
TCA1208-500ML TCA1208-25ML
Cyclohexyl acrylate ≥98.0% stabilized
Cyclohexyl acrylate

(stabilized with MEHQ)
CAS Number: 3066-71-5
MDL Number: MFCD00046349
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 68
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Cyclohexyl Ester

Formula: C₉H₁₄O₂
Boiling Pt: 184 °C
Density: 0.98 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00046349
CAS Number: 3066-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a70ecdc25b4d51562a5ad227eb67d7b1fc33254&vid=6a70ecdc25b4d51562a5ad227eb67d7b1fc33254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR