(3S)-(+)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine
A1174-25G
A1174-10G
TCA1174-25GEA
191.4
USD
TCA1174-25G
TCA1174-010G
(3S)-(+)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
CAS Number: 114715-38-7
MDL Number: MFCD00082766
Molecular Formula: C11H16N2
Molecular Weight: 176.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: 9.5 deg (C=5, H2O)
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Density: 1.02 (20 °C) |
MDL Number:
MFCD00082766 CAS Number: 114715-38-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...