(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

A1173-10G A1173-25G
TCA1173-010GEA 123 USD
TCA1173-010G TCA1173-25G
(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(-)-1-Benzyl-3-aminopyrrolidine

CAS Number: 114715-39-8
MDL Number: MFCD00082638
Molecular Formula: C11H16N2
Molecular Weight: 176.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 279
Flash Point (°C): 140
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: -9.5 deg (C=5, H2O)

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Boiling Pt: 279 °C
Density: 1.02 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00082638
CAS Number: 114715-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db7b84e01dc48f3cdbb69488e7cab03bd8cb4f34&vid=db7b84e01dc48f3cdbb69488e7cab03bd8cb4f34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR