4-Phenylbutylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1123-5ML A1123-25ML
TCA1123-005MLEA 40.28 USD
TCA1123-005ML TCA1123-025ML
4-Phenylbutylamine ≥98.0% (by GC, titration analysis)
4-Phenylbutylamine

CAS Number: 13214-66-9
MDL Number: MFCD00008231
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 101
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1-Amino-4-phenylbutane

Formula: C₁₀H₁₅N
Boiling Pt: 144 °C
Density: 0.94 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00008231
CAS Number: 13214-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=417c23dd83314e36f424306148699f1a3ac4b162&vid=417c23dd83314e36f424306148699f1a3ac4b162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR