Methyl arachidate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl Arachidate
, Methyl eicosanoate
CAS Number: 1120-28-1
MDL Number: MFCD00009014
Molecular Formula: C21H42O2
Molecular Weight: 326.57
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 369
Melting point (°C): 48
Flash Point (°C): 110
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Arachidic Acid Methyl Ester, Eicosanoic Acid Methyl Ester, Icosanoic Acid Methyl Ester, Methyl Eicosanoate, Methyl Icosanoate
Formula:
C₂₁H₄₂O₂ MW: 326.56 g/mol Boiling Pt: 369 °C Melting Pt: 48 °C Flash Pt: 110 °C |
MDL Number:
MFCD00009014 CAS Number: 1120-28-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...