3-(2-Aminoethylamino)propylmethyldimethoxysilane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-(2-Aminoethylamino)propyldimethoxymethylsilane
A0876-500ML
A0876-25ML
TCA0876-500MLEA
261
USD
TCA0876-500ML
TCA0876-25ML
3-(2-Aminoethylamino)propylmethyldimethoxysilane ≥97.0% (by GC, titration analysis)
3-(2-Aminoethylamino)propylmethyldimethoxysilane
CAS Number: 3069-29-2
MDL Number: MFCD00054776
Molecular Formula: C8H22N2O2Si
Molecular Weight: 206.36
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 126
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: N-(2-Aminoethyl)-3-aminopropylmethyldimethoxysilane, N-[3-(Dimethoxymethylsilyl)propyl]ethylenediamine
Formula:
C₈H₂₂N₂O₂Si MW: 206.36 g/mol Boiling Pt: 139 °C Density: 0.97 (20 °C) Flash Pt: 126 °C |
MDL Number:
MFCD00054776 CAS Number: 3069-29-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...