3-Aminopropionitrile fumarate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0796-25G A0796-500G
TCA0796-025GEA 97 USD
TCA0796-025G TCA0796-500G
3-Aminopropionitrile fumarate ≥98.0% (by titrimetric analysis)
3-Aminopropionitrile fumarate

CAS Number: 2079-89-2
MDL Number: MFCD00078321
Molecular Formula: C3H6N2
Molecular Weight: 256.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₀H₁₆N₄O₄
MW: 256.26 g/mol
MDL Number: MFCD00013142
CAS Number: 2079-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd0f74426efac3f245ba85d31012d6ab5e66a628&vid=29bba65eb8740dbb146ae705a2fb0f777cbaa24e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR