Isobutyl acrylate ≥99.0% stabilized

Supplier: TCI America

A0747-500ML A0747-25ML
TCA0747-500MLEA 61.44 USD
TCA0747-500ML TCA0747-25ML
Isobutyl acrylate ≥99.0% stabilized
Isobutyl acrylate

(stabilized with MEHQ)
CAS Number: 106-63-8
MDL Number: MFCD00042865
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 31
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Isobutyl Ester

Formula: C₇H₁₂O₂
Boiling Pt: 132 °C
Density: 0.89 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00042865
CAS Number: 106-63-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=812af4e19bed9c335c9c6fa9f765d2b5b16023c8&vid=812af4e19bed9c335c9c6fa9f765d2b5b16023c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR