1-Adamantaneacetic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane

A0736-5G A0736-25G
TCA0736-005GEA 74 USD
TCA0736-005G TCA0736-025G
1-Adamantaneacetic acid ≥99.0% (by titrimetric analysis)
1-Adamantaneacetic acid

CAS Number: 4942-47-6
MDL Number: MFCD00074728
Molecular Formula: C12H18O2
Molecular Weight: 194.27
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 1-Carboxymethyladamantane

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00074728
CAS Number: 4942-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac96896487da266f754e40317a0bc3f94ae0cc8&vid=bac96896487da266f754e40317a0bc3f94ae0cc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR