Arbutin ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside
CAS Number: 497-76-7
MDL Number: MFCD00016915
Molecular Formula: C12H16O7
Molecular Weight: 272.25
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 198
Specific rotation [a]20/D: -65.5 deg (C=4, H2O)
Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Cosmetic Ingredients for Research and Experimental Use, Others (Phytochemicals)
Other Product Names / Synonyms: Hydroquinone-beta-D-glucoside, 4-Hydroxyphenyl beta-D-Glucopyranoside
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00016915 CAS Number: 497-76-7 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (HPLC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...