Masterflex® Transfer Tubing, Flexible PVC, 3/8" ID x 9/16" OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX95505-28
Your Organization's Price:
Retrieving
As flexible as you need it.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: ³/₈" int.Ø × ⁹/₁₆" out.Ø
<B>Applications:</B> Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications.<br /><B>Characteristics:</B>Nontoxic DEHP-free formulation. Flexible, small bend radii. Transparent; allows for visual inspection and flow monitoring.<br /> <B>Certifications:</B> FDA compliant, Proposition 65, REACH<br /><B>Temperature range:</B> -58 to 165 °F (-50 to 73 °C)<br /> <B>Sterilize:</B> Autoclave.