Masterflex® Ismatec® Pump Tubing, Solva™ Solvent/Hydrocarbon, 0.38 mm ID; 50 ft
Supplier: VWR International
VWR Item#MFLX95712-14
Your Organization's Price:
Retrieving
Specifically designed for use with hydrocarbons, petroleum products and distillates.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Resists most hydrocarbons, embrittlement, and swelling
  • Tubing Inner Diameter: 0.38 mm
Resistant to ozone and UV light — won't crack, swell, or harden.<br />Translucent yellow color<br />Static temperature range: -37 to 74 °C (-35 to 165 °F)<br />Dynamic temperature range: 0 to 40 °C (32 to 104 °F)<br /><br />The overall length of Solvent/Hydrocarbon extension tubing is 50 feet (15 m). The legacy Ismatec® extension tubing was 3 m long.