Masterflex® Microbore Transfer Tubing, Tygon® ND-100-80, 0.020" ID x 0.060" OD; 100 Ft
Supplier: VWR International
VWR Item#MFLX06419-01
Your Organization's Price:
Retrieving
Clear formulation for easier sample viewing.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 0.020" int.Ø × 0.060" out.Ø
Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements.
Temperature range: -25 to 185 °F (-31 to 85 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.