Masterflex® L/S® Replacement Fuse, Slo-Blo, 2.5 A/250 V, 5 X 20 mm
Supplier: Avantor Fluid Handling
Make the most of your drive with these replacement parts.
- Optimize performance of your Masterflex pumps
Suitable for use with Masterflex® B/T® drive 77111-30.
For use with the following Masterflex I/P drives: 07591-05, 07591-07, 07591-15, 07591-65, 07591-67, and 07591-75; 77310-00 and 77310-01; 77340-00.