Blue-White® Variable-Area Flowmeter, In-Line, Direct-Read, Polysulfone Housing, 0.25 GPM Water
Supplier: VWR International
VWR Item#MFLX32470-00
Your Organization's Price:
Retrieving
Get better results than acrylic.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Choose from standard, high-capacity, or ultrapure in-line flowmeters. Standard models are designed for low flow readings—measure flow rates as low as 0.025 GPM. The high-capacity flowmeters measure flow rate as high as 130 GPM! <br /><br />These large, durable flowmeters have 316 SS internal parts. New ultrapure flowmeters use only plastic wetted parts, making these great for ultrapure water such as deionized water. For high-capacity models with polycarbonate shields around the flowtube, call our Application Specialists and get more information.