Masterflex® Fitting, Polypropylene, Straight, Male Luer to Hosebarb Adapter, 3/32" ID; 25/PK
Supplier: VWR International
VWR Item#MFLX30800-28
Your Organization's Price:
Retrieving
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male luer to ³/₃₂" hose barb with 0.068" diameter orifice