Masterflex® Fitting, Polypropylene, Straight, Male Luer to Hosebarb Adapter, 3/16" ID; 25/PK
Supplier: VWR International
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Male luer to ³/₁₆" hose barb