Sanitary Plastic Ball Valve, 2-Way, 3/4" Mini Tri-Clamp, Polypropylene, EPDM Seal
Supplier: VWR International
VWR Item#MFLX30532-01
Your Organization's Price:
Retrieving
Precision machined and tested to ensure leak-free connections.
  • Standard Tri-Clamp connections are easy to plumb into sanitary fluid paths
  • Smooth flow path minimizes turbulence and pressure loss
  • Valve design permits full flow and pressure in either direction
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.