VWR® Atlas Slot-Mounted Shelving, Double-Sided
Supplier: VWR International
VWRDSNL1236 VWRDSNL1242 VWRDSNL1248 VWRDSNL1254 VWRDSNL1260 VWRDSNL1266 VWRDSNL1272 VWRDSNL1536 VWRDSNL1542 VWRDSNL1548 VWRDSNL1554 VWRDSNL1560 VWRDSNL1566 VWRDSNL1572 VWRDSNL1836 VWRDSNL1842 VWRDSNL1848 VWRDSNL1854 VWRDSNL1860 VWRDSNL1866 VWRDSNL1872 VWRDSNL1560-W
VWRDSNL1236EA 398.84 USD
VWRDSNL1236 VWRDSNL1242 VWRDSNL1248 VWRDSNL1254 VWRDSNL1260 VWRDSNL1266 VWRDSNL1272 VWRDSNL1536 VWRDSNL1542 VWRDSNL1548 VWRDSNL1554 VWRDSNL1560 VWRDSNL1566 VWRDSNL1572 VWRDSNL1836 VWRDSNL1842 VWRDSNL1848 VWRDSNL1854 VWRDSNL1860 VWRDSNL1866 VWRDSNL1872 VWRDSNL1560-W
VWR® Atlas Slot-Mounted Shelving, Double-Sided
Furniture Shelves
Slot-mounted shelving for double-sided applications.
  • Adjustable with use of uprights
Used with slotted uprights (see VWRSU24 series) for shelf adjustment in 2.5 cm (1") increments.
Información para pedidos: Two shelves are required; each is sold separately. Brackets are included.
Order Now
SPECIFICATIONS
Environmentally Preferable