REDISHIP Supreme Air® Fittings for KMER-Lined Bench and Walk-In Hoods, Kewaunee®
Supplier: VWR International
L3185NPHLRPA L3185NPHLRPG L3185NPHLRPN L3185NPHLRPV L3185WPHLRP L3185WPHLRTL L3185WPH9RSVB
VFT-0860-3AEA 124.35 USD
VFT-0860-3A VFT-0860-3G VFT-0860-3N VFT-0860-3V VFT-0861-3W VFT-0862-3D VFT-0864-3WV
REDISHIP Supreme Air® Fittings for KMER-Lined Bench and Walk-In Hoods, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
These remote control fittings consist of a brass angle valve with replaceable seat, a nylon color-coded panel flange with angle serrated hose connector (except VFT-0864-3WV, which features a gooseneck), a remote control rod assembly with a plastic color-coded four-arm handle, black plastic angle flange, and a tank nipple, locknut, and washer
Fittings may be used on either side of the hood. Not for use on Supreme Air Isotope hoods.
Información para pedidos: REDISHIP units can be shipped within 48 hours of receiving your order.
Order Now
SPECIFICATIONS
Environmentally Preferable