CleanTips® Large Rectanglar Head Foam Swab, Texwipe®
Supplier: Texwipe
TX712A
TWTX712ACS 617.09 USD
TWTX712A
CleanTips® Large Rectanglar Head Foam Swab, Texwipe®
Swabs Swabs for Cleaning
The rectangular head swab is a large, general-purpose swab for cleaning surfaces and hard-to-reach areas
Swab features a rectangular foam head on a rigid internal paddle for firm support. Made from closed-cell polyurethane foam with 100 pore-per-inch porosity, this swab possesses excellent durability and cushioning. Thermally sealed head eliminates need for adhesives. High-grade polyurethane foam head will not crack or chip. Low in fiber and particle generation. Good chemical resistance.
The large, light green polypropylene handle enables good control and ease of use.