Temozolomide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide

46676-0010 466760050
TS46676-0010EA 150 USD
TS46676-0010 TS466760050
Temozolomide
Temozolomide
Formula: C₆H₆N₆O₂
MW: 194.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00866492
CAS Number: 85622-93-1
Merck Index: 13.9218

Order Now

Specification Test Results

Appearance White to light pink to brown powder
Infrared spectrum Conforms
Loss on drying ≤ 1%
HPLC ≥ 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR