Theophylline-7-acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid

45925-1000
TS45925-0250EA 29.87 USD
TS45925-0250 TS45925-1000 TS45925-5000
Theophylline-7-acetic acid 98%
Theophylline-7-acetic acid
Formula: C₉H₁₀N₄O₄
MW: 238.20 g/mol
Melting Pt: 270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022832
CAS Number: 652-37-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 268 to 272 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR