(R)-3-(Aminomethyl)-1-boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1-BOC-3-(Aminomethyl)piperidine

43642-0010 43642-0050
TS43642-0010EA 139 USD
TS43642-0010 TS43642-0050
(R)-3-(Aminomethyl)-1-boc-piperidine 97%
(R)-3-(Aminomethyl)-1-Boc-piperidine
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03839876
CAS Number: 140645-23-4

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +10° to +20° (20°C, 589 nm) (c=1 in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR