Petrolatum, white, pure

Supplier: Thermo Scientific Chemicals

41709-0010 417090100
TS41709-0010EA 51 USD
TS41709-0010 TS417090100
Petrolatum, white, pure
Petrolatum
Boiling Pt: 300 °C
Melting Pt: 50…56 °C
Density: 0.84 g/cm³
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00147839
CAS Number: 8009-03-8

Order Now

Specification Test Results

Appearance White Paste
Infrared spectrum Conforms
Viscosity 5 to 8 mm²/s (at 100 °C)
Solidification point 50 to 56°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR