Indocyanine green, pure laser grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen

41254-0020 41254-1000 41254-5000
TS41254-0020EA 2033 USD
TS41254-0020 TS41254-1000 TS41254-5000
Indocyanine green, pure laser grade
Indocyanine green
Formula: C₄₃H₄₇N₂NaO₆S₂
MW: 774.99
Melting Pt: 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013078
CAS Number: 3599-32-4

Order Now

Specification Test Results

Appearance Green to brown Crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 784 nm (in Methanol)
Molar absorption coefficient ≥230000 l/(mol.cm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR