1-Heptanesulfonic acid sodium salt monohydrate for HPLC

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 1-heptanesulphonate monohydrate

41127-0250 41127-1000 AC41127-0050 AC41127-5000
TS41127-0250EA 208 USD
TS41127-0250 TS41127-1000 TS41127-0050 TS41127-5000
1-Heptanesulfonic acid sodium salt monohydrate for HPLC
1-Heptanesulphonic acid sodium salt monohydrate
Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5

Order Now

Specification Test Results

Appearance White Powder or flakes
Infrared spectrum Conforms
Loss on drying 7.4 % to 9 % (100°C, 3 hrs) (vacuum)
Solubility (10 % w/v in water) Clear colorless
UV at 260 nm A: ≤0.01
Titration after Ion exchange ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR