2,4-Dibromophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

40651-0050 40651-1000
TS40651-0050EA 25 USD
TS40651-0050 TS40651-1000
2,4-Dibromophenol 99%
2,4-Dibromophenol
Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 154 °C (11 mmHg)
Melting Pt: 38
Flash Pt: >110 °C
MDL Number: MFCD00002149
CAS Number: 615-58-7
EINECS: 210-436-5
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
Melting point 36 to 39 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR